α,α,α,2-Tetrafluorotoluene ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A18095.06
A18095.14
A18095.22
A18095.06EA
924
CZK
A18095.06
A18095.14
A18095.22
α,α,α,2-Tetrafluorotoluene ≥99%
α,α,α,2-Tetrafluorotoluene
Purity: 99%
CAS No.: 392-85-8
Molecular Formula: C7H4F4
Formula Weight: 164.10
Flash Point: 17°(62°F)
Refractive Index: 1.4060
Melting Point: -49°
Boiling Point: 114-115°
Beilstein Reference: 2046638
UN No.: 1993
MDL No.: MFCD00000374
Vzorec:
C₇H₄F₄ Bod varu: 114…115 °C Bod tání: –49 °C Hustota: 1,293 Bod vzplanutí: 17 °C (62 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000374 CAS číslo: 392-85-8 EINECS: 206-880-4 UN: 1993 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...