Tisk…

2,4-Difluorotoluene ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A11667.06 A11667.14
A11667.06EA 642 CZK
A11667.06 A11667.14
2,4-Difluorotoluene ≥98%
2,4-Difluorotoluene

Purity: 98+% CAS No.: 452-76-6 Molecular Formula: C7H6F2 Formula Weight: 128.12 Flash Point: 15°(59°F) Refractive Index: 1.4490 Odour: Aromatic Boiling Point: 114-115° Beilstein Reference: 1931681 UN No.: 2388 MDL No.: MFCD00000332

Vzorec: C₇H₆F₂
Bod varu: 114…115 °C
Hustota: 1,121
Bod vzplanutí: 15 °C (59 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000332
CAS číslo: 452-76-6
EINECS: 207-211-9
UN: 2388
ADR: 3,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR