2,6-Difluorotoluene ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
1,3-Difluoro-2-methylbenzene
B25076.06
B25076.14
B25076.06EA
1150
CZK
B25076.06
B25076.14
2,6-Difluorotoluene ≥98%
2,6-Difluorotoluene
Purity: 98%
CAS No.: 443-84-5
Molecular Formula: C7H6F2
Formula Weight: 128.12
Flash Point: 10°(50°F)
Refractive Index: 1.4528
Odour: Aromatic
Boiling Point: 112-115°
Beilstein Reference: 1932656
UN No.: 2388
MDL No.: MFCD00043898
Vzorec:
C₇H₆F₂ Bod varu: 112…115 °C Hustota: 1,130 Bod vzplanutí: 10 °C (50 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00043898 CAS číslo: 443-84-5 UN: 2388 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...