Tisk…

α-Chloro-3,5-dinitrotoluene, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

42036-25G 42036-5G
SIAL42036-25GEA 12140 CZK
SIAL42036-25G SIAL42036-5G
α-Chloro-3,5-dinitrotoluene, Sigma-Aldrich®
α-Chloro-3,5-dinitrotoluene
Vzorec: C₇H₅ClN₂O₄
Storage Temperature: Ambient
CAS číslo: 74367-78-5
EINECS: 277-841-7
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR