Tisk…

4-Chloro-3-nitrotoluene 97+%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 1-Chloro-4-methyl-2-nitrobenzene

A11383.22 A11383.36
A11383.22EA 2660 CZK
A11383.22 A11383.36
4-Chloro-3-nitrotoluene 97+%
4-Chloro-3-nitrotoluene

Purity: 97+% CAS No.: 89-60-1 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Flash Point: 129°(264°F) Refractive Index: 1.5580 Melting Point: ca 7° Boiling Point: 260-262° Beilstein Reference: 511055 UN No.: 2433 MDL No.: MFCD00007085

Vzorec: C₇H₆ClNO₂
MW: 171,58 g/mol
Bod varu: 260…262 °C
Bod tání: ∼7 °C
Hustota: 1,295
Bod vzplanutí: 129 °C (264 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007085
CAS číslo: 89-60-1
EINECS: 201-922-8
UN: 2433
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR