4-Chloro-α,α,α-trifluorotoluene ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
1-Chloro-4-(trifluoromethyl)benzene, 4-Chlorobenzotrifluoride
A15154.0E
A15154.22
A15154.36
A15154.0EEA
8580
CZK
A15154.0E
A15154.22
A15154.36
4-Chloro-α,α,α-trifluorotoluene ≥98%
4-Chloro-α,α,α-trifluorotoluene
Purity: 98+%
CAS No.: 98-56-6
Molecular Formula: C7H4ClF3
Formula Weight: 180.56
Flash Point: 47°(116°F)
Refractive Index: 1.4460
Melting Point: -36°
Boiling Point: 136-138°
Beilstein Reference: 510203
Merck Reference: 14,2126
UN No.: 2234
MDL No.: MFCD00000627
Vzorec:
C₇H₄ClF₃ Bod varu: 136…138 °C Bod tání: –36 °C Hustota: 1,346 Bod vzplanutí: 47 °C (116 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000627 CAS číslo: 98-56-6 EINECS: 202-681-1 UN: 2234 ADR: 3,III Merck Index: 14,2126 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...