α-Chloro-3-fluorotoulene 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
1-(Chloromethyl)-3-fluorobenzene, α-Chloro-m-fluorotoulene, 3-Fluorobenzyl chloride
A12665.14
A12665.06
A12665.22
A12665.14EA
4090
CZK
A12665.14
A12665.06
A12665.22
α-Chloro-3-fluorotoulene 97%
α-Chloro-3-fluorotoulene
Purity: 97%
CAS No.: 456-42-8
Molecular Formula: C7H6ClF
Formula Weight: 144.58
Flash Point: 58°(136°F)
Refractive Index: 1.5120
Odour: Pungent
Boiling Point: 176-177°
Beilstein Reference: 742265
UN No.: 2920
MDL No.: MFCD00000904
Vzorec:
C₇H₆ClF MW: 144,58 g/mol Bod varu: 176…177 °C Hustota: 1,213 Bod vzplanutí: 58 °C (136 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000904 CAS číslo: 456-42-8 EINECS: 207-264-8 UN: 2920 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...