α-Chloro-2-fluorotoulene ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-Fluorobenzyl chloride, 1-(Chloromethyl)-2-fluorobenzene, α-Chloro-o-fluorotoulene
A11566.14
A11566.22
A11566.14EA
1660
CZK
A11566.14
A11566.22
α-Chloro-2-fluorotoulene ≥98%
α-Chloro-2-fluorotoulene
Purity: 98+%
CAS No.: 345-35-7
Molecular Formula: C7H6ClF
Formula Weight: 144.58
Flash Point: 57°(134°F)
Refractive Index: 1.5154
Odour: Pungent
Boiling Point: 175°
Beilstein Reference: 471699
UN No.: 2920
MDL No.: MFCD00000892
Vzorec:
C₇H₆ClF MW: 144,58 g/mol Bod varu: 175 °C Hustota: 1,216 Bod vzplanutí: 57 °C (134 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000892 CAS číslo: 345-35-7 EINECS: 206-460-0 UN: 2920 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...