Tisk…

α,4-Dichlorotoluene ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 1-Chloro-4-(chloromethyl)benzene, 4-Chlorobenzyl chloride

A14696.0B A14696.0I A14696.14 A14696.30
A14696.0BEA 4810 CZK
A14696.0B A14696.0I A14696.14 A14696.30
α,4-Dichlorotoluene ≥98%
α,4-Dichlorotoluene

Purity: 98+% CAS No.: 104-83-6 Molecular Formula: C7H6Cl2 Formula Weight: 161.03 Flash Point: 107°(225°F) Refractive Index: 1.5575 Odour: Pungent Melting Point: 26-30° Boiling Point: 220-221° Beilstein Reference: 471558 UN No.: 3427 MDL No.: MFCD00000914

Vzorec: C₇H₆Cl₂
MW: 161,03 g/mol
Bod varu: 220…221 °C
Bod tání: 26…30 °C
Hustota: 1,275
Bod vzplanutí: 107 °C (225 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000914
CAS číslo: 104-83-6
EINECS: 203-242-7
UN: 3427
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR