Tisk…

α,2,5-Trichlorotoluene ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 2,5-Dichlorobenzyl chloride, 1,4-Dichloro-2-(chloromethyl)benzene, 1-(Chloromethyl)-2,5-dichlorobenzene

L07506.06 L07506.14
L07506.06EA 2810 CZK
L07506.06 L07506.14
α,2,5-Trichlorotoluene ≥97%
α,2,5-Trichlorotoluene

Purity: 97% CAS No.: 2745-49-5 Molecular Formula: C7H5Cl3 Formula Weight: 195.48 Refractive Index: 1.5755 Odour: Pungent Boiling Point: 121-124°/14mm Beilstein Reference: 637802 UN No.: 3265 MDL No.: MFCD00035818

Vzorec: C₇H₅Cl₃
MW: 195,47 g/mol
Bod varu: 121…124 °C (14 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00035818
CAS číslo: 2745-49-5
EINECS: 220-382-4
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR