2,3-Dichlorotoluene ≥98%
Dodavatel: Thermo Fisher Scientific
Varování
|
A11206.14
A11206.18
A11206.22
A11206.14EA
2510
CZK
A11206.14
A11206.18
A11206.22
2,3-Dichlorotoluene ≥98%
2,3-Dichlorotoluene
Purity: 98%
CAS No.: 32768-54-0
Molecular Formula: C7H6Cl2
Formula Weight: 161.03
Flash Point: 83°(181°F)
Refractive Index: 1.5510
Melting Point: 4°
Boiling Point: 207-208°
Beilstein Reference: 2412384
UN No.: 3082
MDL No.: MFCD00000546
Vzorec:
C₇H₆Cl₂ Bod varu: 207…208 °C Bod tání: 4 °C Hustota: 1,229 Bod vzplanutí: 83 °C (181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000546 CAS číslo: 32768-54-0 EINECS: 251-203-8 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...