Benzotrichloride 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
α,α,α-Trichlorotoluene, Phenylchloroform, (Trichloromethyl)benzene
164530010
164530050
ACRO164530010EA
7030
CZK
ACRO164530010
ACRO164530050
Benzotrichloride 98%
Benzotrichloride
Vzorec:
C₇H₅Cl₃ MW: 195,47 g/mol Bod varu: 220,6 °C (1013 hPa) Bod tání: –7,5 °C Hustota: 1,38 g/cm³ Bod vzplanutí: 97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000786 CAS číslo: 98-07-7 EINECS: 202-634-5 UN: 2226 ADR: 8,II Merck Index: 13,01110 |
Výsledky specifikačního testu
Appearance | Clear yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5570 to 1.5590 (20°C, 589 nm) |
GC | ≥97.5 % |
Chloride (Cl) | ≤0.5 ppm |
Nitrate (NO3) | ≤0.2 ppm |
Aluminium (Al) | ≤0.05 ppm |
Ammonium (NH4) | ≤2 ppm |
Arsenic (As) | ≤0.01 ppm |
Beryllium (Be) | ≤0.02 ppm |
Cadmium (Cd) | ≤0.02 ppm |
Calcium (Ca) | ≤0.2 ppm |
Chromium (Cr) | ≤0.05 ppm |
Cobalt (Co) | ≤0.02 ppm |
Copper (Cu) | ≤0.01 ppm |
Iron (Fe) | ≤0.1 ppm |
Lead (Pb) | ≤0.02 ppm |
Lithium (Li) | ≤0.02 ppm |
Magnesium (Mg) | ≤0.05 ppm |
Manganese (Mn) | ≤0.01 ppm |
Nickel (Ni) | ≤0.02 ppm |
Potassium (K) | ≤0.1 ppm |
Silver (Ag) | ≤0.02 ppm |
Sodium (Na) | ≤0.5 ppm |
Strontium (Sr) | ≤0.02 ppm |
Zinc (Zn) | ≤0.1 ppm |
Heavy metals (as Pb) | ≤1 ppm |
KMnO4-reducing substance | ≤0.0002 % (as SO2) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...