Tisk…

6-Hydroxy-m-toluic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 5-Methylsalicylic acid, 2,5-Cresotic acid

B20040.06 B20040.14
B20040.06EA 1440 CZK
B20040.06 B20040.14
6-Hydroxy-m-toluic acid ≥98%
6-Hydroxy-m-toluic acid

Purity: 98% CAS No.: 89-56-5 Molecular Formula: C8H8O3 Formula Weight: 152.15 Odour: Odourless Melting Point: 151-154° MDL No.: MFCD00002461

Vzorec: C₈H₈O₃
MW: 152.15
Bod varu: 313
Bod tání: 151…154 °C
Hustota: 1.31 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002461
CAS číslo: 89-56-5
EINECS: 201-918-6
Merck Index: 12,02650

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR