2-Hydroxy-m-toluic acid ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
2,3-Cresotic acid, 2-Hydroxy-3-methylbenzoic acid, 3-Methylsalicylic acid
L15825.14
L15825.22
L15825.14EA
604
CZK
L15825.14
L15825.22
2-Hydroxy-m-toluic acid ≥99%
2-Hydroxy-m-toluic acid
Purity: 99%
CAS No.: 83-40-9
Molecular Formula: C8H8O3
Formula Weight: 152.15
Odour: Odourless
Melting Point: 163-167°
Beilstein Reference: 2086811
Merck Reference: 14,2581
MDL No.: MFCD00002448
Vzorec:
C₈H₈O₃ MW: 152.15 Bod varu: 293 Bod tání: 163…167 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002448 CAS číslo: 83-40-9 EINECS: 201-473-8 Merck Index: 13,02609 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...