Tisk…

p-Toluic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 4-Methylbenzoic acid, 4-Toluic acid

A11506.0B A11506.0I A11506.30
A11506.0BEA 3420 CZK
A11506.0B A11506.0I A11506.30
p-Toluic acid ≥98%
p-Toluic acid

Purity: 98% CAS No.: 99-94-5 Molecular Formula: C8H8O2 Formula Weight: 136.15 Flash Point: 181°(357°F) Melting Point: 178-182° Boiling Point: 274° Beilstein Reference: 507600 Merck Reference: 14,9535 MDL No.: MFCD00002565

Vzorec: C₈H₈O₂
MW: 136.15
Bod varu: 274 °C
Bod tání: 178…182 °C
Hustota: 1.06
Bod vzplanutí: 181 °C (357 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002565
CAS číslo: 99-94-5
EINECS: 202-803-3
Merck Index: 12,09673

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR