4-Heptylbenzoic acid ≥99%
Dodavatel: Thermo Fisher Scientific
B22589.06
B22589.14
B22589.06EA
1680
CZK
B22589.06
B22589.14
4-Heptylbenzoic acid ≥99%
4-Heptylbenzoic acid
Purity: 99+%
CAS No.: 38350-87-7
Molecular Formula: C14H20O2
Formula Weight: 220.31
Melting Point: 101-102°
Beilstein Reference: 2047135
Vzorec:
C₁₄H₂₀O₂ |
MDL Number:
MFCD00009722 CAS číslo: 38350-87-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...