Tisk…

4-Ethylbenzoic acid 97%

Dodavatel: Thermo Scientific

184490050 184490250
ACRO184490050EA 478 CZK
ACRO184490050 ACRO184490250
4-Ethylbenzoic acid 97%
4-Ethylbenzoic acid
Vzorec: C₉H₁₀O₂
MW: 150,18 g/mol
MDL Number: MFCD00002570
CAS číslo: 619-64-7

Objednat

Výsledky specifikačního testu

Appearance White to beige Powder
Infrared spectrum Conforms
Melting point 110 to 114 °C
Titration with TBAH ≥96.0 %
pH 11 to 12 (1 % in water)
Specific optical rotation -9° to -13° (20 °C, 589 nm) (4 % in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR