4-tert-Butylbenzoic acid ≥99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A13997.30
A13997.0B
A13997.0I
A13997.30EA
841
CZK
A13997.30
A13997.0B
A13997.0I
4-tert-Butylbenzoic acid ≥99%
4-tert-Butylbenzoic acid
Purity: 99+%
CAS No.: 98-73-7
Molecular Formula: C11H14O2
Formula Weight: 178.23
Flash Point: 180°(356°F)
Melting Point: 164-168°
Boiling Point: 280° dec.
Beilstein Reference: 607545
UN No.: 2811
MDL No.: MFCD00002563
Vzorec:
C₁₁H₁₄O₂ Bod varu: 280 °C (rozkl.) Bod tání: 164…168 °C Hustota: 0,6 g/cm³ (20 °C) Bod vzplanutí: 180 °C (356 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002563 CAS číslo: 98-73-7 EINECS: 202-696-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...