Valerophenone ≥99%
Dodavatel: Thermo Scientific
A10525.14
A10525.22
A10525.36
A10525.14EA
1500
CZK
A10525.14
A10525.22
A10525.36
Valerophenone ≥99%
Valerophenone
Purity: 99%
CAS No.: 1009-14-9
Molecular Formula: C11H14O
Formula Weight: 162.23
Flash Point: 102°(215°F)
Refractive Index: 1.5143
Melting Point: -9°
Boiling Point: 244-245°
Beilstein Reference: 1907717
MDL No.: MFCD00009480
Vzorec:
C₆H₅C(O)(CH₂)₃CH₃ Bod varu: 244…245 °C Bod tání: –9 °C Hustota: 0,978 Bod vzplanutí: 102 °C (215 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009480 CAS číslo: 1009-14-9 EINECS: 213-767-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...