Tisk…

Octanophenone 98%

Dodavatel: Thermo Scientific

Synonyms: n-Octanophenone

A13652.06 A13652.14
A13652.06EA 2130 CZK
A13652.06 A13652.14
Octanophenone 98%
Octanophenone

Purity: 98% CAS No.: 1674-37-9 Molecular Formula: C14H20O Formula Weight: 204.31 Flash Point: 110°(230°F) Refractive Index: 1.5040 Melting Point: 22-24° Boiling Point: 285° Beilstein Reference: 1868534 MDL No.: MFCD00003554

Vzorec: C₆H₅C(O)(CH₂)₆CH₃
MW: 204,31 g/mol
Bod varu: 285 °C
Bod tání: 22…24 °C
Hustota: 0,938
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003554
CAS číslo: 1674-37-9
EINECS: 216-817-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR