Octanophenone 99+%
Dodavatel: Thermo Fisher Scientific
279360250
279360050
279361000
ACRO279360250EA
6590
CZK
ACRO279360250
ACRO279360050
ACRO279361000
Octanophenone 99+%
Octanophenone
Vzorec:
C₆H₅C(O)(CH₂)₆CH₃ Bod varu: 285…290 °C (760 mmHg) Hustota: 0,93 g/cm³ Bod vzplanutí: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003554 CAS číslo: 1674-37-9 EINECS: 216-817-2 |
|
Výsledky specifikačního testu
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5024 to 1.5054 (20°C, 589 nm) |
GC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...