Tisk…

4-tert-Butylbenzonitrile ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: p-tert-Butylbenzonitrile, 4-(tert-Butyl)benzonitrile

A14076.03 A14076.06 A14076.14
A14076.03EA 900 CZK
A14076.03 A14076.06 A14076.14
4-tert-Butylbenzonitrile ≥98%
4-tert-Butylbenzonitrile

Purity: 98+% CAS No.: 4210-32-6 Molecular Formula: C11H13N Formula Weight: 159.23 Flash Point: 110°(230°F) Refractive Index: 1.5170 Odour: Characteristic Boiling Point: 258° Beilstein Reference: 2080049 MDL No.: MFCD00075840

Vzorec: C₁₁H₁₃N
MW: 159,23 g/mol
Bod varu: 258 °C
Hustota: 0,940
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00075840
CAS číslo: 4210-32-6
EINECS: 224-137-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR