4-Isopropylbenzaldehyde tech. 90%, technická kvalita
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A14019.22
A14019.14
A14019.36
A14019.22EA
1370
CZK
A14019.22
A14019.14
A14019.36
4-Isopropylbenzaldehyde tech. 90%, technická kvalita
4-Isopropylbenzaldehyde
Purity: 90%
CAS No.: 122-03-2
Molecular Formula: C10H12O
Formula Weight: 148.21
Flash Point: 93°(199°F)
Refractive Index: 1.5300
Boiling Point: 234-235°
Beilstein Reference: 636547
Merck Reference: 14,2621
MDL No.: MFCD00006953
Vzorec:
C₁₀H₁₂O Bod varu: 234…235 °C Hustota: 0,979 Bod vzplanutí: 93 °C (199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006953 CAS číslo: 122-03-2 EINECS: 204-516-9 Merck Index: 14,2621 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...