4-tert-Butylbenzaldehyde ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
4-(tert-Butyl)benzaldehyde
A15740.14
A15740.06
A15740.22
A15740.14EA
5360
CZK
A15740.14
A15740.06
A15740.22
4-tert-Butylbenzaldehyde ≥97%
4-tert-Butylbenzaldehyde
Purity: 97%
CAS No.: 939-97-9
Molecular Formula: C11H14O
Formula Weight: 162.23
Flash Point: 101°(213°F)
Refractive Index: 1.5285
Boiling Point: 244-246°
Beilstein Reference: 1906461
UN No.: 3082
MDL No.: MFCD00035742
Vzorec:
C₁₁H₁₄O MW: 162,23 g/mol Bod varu: 244…246 °C Hustota: 0,970 Bod vzplanutí: 101 °C (213 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00035742 CAS číslo: 939-97-9 EINECS: 213-367-9 UN: 3082 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...