4-Bromo-2-nitrotoluene ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
A12951.06
A12951.22
A12951.06EA
1490
CZK
A12951.06
A12951.22
4-Bromo-2-nitrotoluene ≥99%
4-Bromo-2-nitrotoluene
Purity: 99%
CAS No.: 60956-26-5
Molecular Formula: C7H6BrNO2
Formula Weight: 216.04
Flash Point:
Vzorec:
C₇H₆BrNO₂ MW: 216,03 g/mol Bod varu: 130 °C (12 mmHg) Bod tání: 45…48 °C Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041243 CAS číslo: 60956-26-5 EINECS: 262-536-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...