Tisk…

2-Bromo-4-nitrotoluene ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

A14865.14 A14865.22
A14865.14EA 1940 CZK
A14865.14 A14865.22
2-Bromo-4-nitrotoluene ≥98%
2-Bromo-4-nitrotoluene

Purity: 98% CAS No.: 7745-93-9 Molecular Formula: C7H6BrNO2 Formula Weight: 216.04 Melting Point: 74-78° Beilstein Reference: 1941232

Vzorec: C₇H₆BrNO₂
MW: 216,03 g/mol
Bod varu: 175 °C (38 mmHg)
Bod tání: 74…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00007195
CAS číslo: 7745-93-9
EINECS: 231-809-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR