α-Bromo-3-nitrotoluene ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A14310.06
A14310.14
A14310.22
A14310.06EA
2050
CZK
A14310.06
A14310.14
A14310.22
α-Bromo-3-nitrotoluene ≥98%
α-Bromo-3-nitrotoluene
Purity: 98+%
CAS No.: 3958-57-4
Molecular Formula: C7H6BrNO2
Formula Weight: 216.04
Melting Point: 56-59°
Boiling Point: 153-154°/7mm
UN No.: 3261
MDL No.: MFCD00007271
Vzorec:
C₇H₆BrNO₂ Bod varu: 153…154 °C (7 mmHg) Bod tání: 56…59 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007271 CAS číslo: 3958-57-4 EINECS: 223-557-3 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...