Tisk…

4-Bromo-α,α,α-trifluorotoluene ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 4-Bromobenzotrifluoride, 1-Bromo-4-(trifluoromethyl)benzene

A15942.06 A15942.14 A15942.22
A15942.06EA 1570 CZK
A15942.06 A15942.14 A15942.22
4-Bromo-α,α,α-trifluorotoluene ≥99%
4-Bromo-α,α,α-trifluorotoluene

Purity: 99% CAS No.: 402-43-7 Molecular Formula: C7H4BrF3 Formula Weight: 225.01 Flash Point: 60°(140°F) Refractive Index: 1.4730 Boiling Point: 154-155° Beilstein Reference: 2045666 UN No.: 1993 MDL No.: MFCD00000398

Vzorec: C₇H₄BrF₃
Bod varu: 154…155 °C
Hustota: 1,630
Bod vzplanutí: 60 °C (140 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000398
CAS číslo: 402-43-7
EINECS: 206-943-6
UN: 1993
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR