Tisk…

2-Bromo-4-fluorotoluene ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 1-Bromo-3-fluoro-6-methylbenzene

A12792.06 A12792.14 A12792.22
A12792.06EA 1240 CZK
A12792.06 A12792.14 A12792.22
2-Bromo-4-fluorotoluene ≥98%
2-Bromo-4-fluorotoluene

Purity: 98+% CAS No.: 1422-53-3 Molecular Formula: C7H6BrF Formula Weight: 189.03 Flash Point: 64°(147°F) Refractive Index: 1.5280 Boiling Point: 170-172° Beilstein Reference: 1859038 MDL No.: MFCD00040828

Vzorec: C₇H₆BrF
MW: 189,03 g/mol
Bod varu: 170…172 °C
Hustota: 1,526
Bod vzplanutí: 64 °C (147 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040828
CAS číslo: 1422-53-3
EINECS: 215-830-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR