Tisk…

α-Bromo-2,6-difluorotoluene ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 2-(Bromomethyl)-1,3-difluorobenzene, 2,6-Difluorobenzyl bromide

A17434.06 A17434.14
A17434.06EA 1940 CZK
A17434.06 A17434.14
α-Bromo-2,6-difluorotoluene ≥97%
α-Bromo-2,6-difluorotoluene

Purity: 97+% CAS No.: 85118-00-9 Molecular Formula: C7H5BrF2 Formula Weight: 207.02 Flash Point: 110°(230°F) Melting Point: 49-53° Beilstein Reference: 2083943 UN No.: 1759 MDL No.: MFCD00000329

Vzorec: C₇H₅BrF₂
MW: 207,02 g/mol
Bod tání: 49…53 °C
Hustota: 1,600
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000329
CAS číslo: 85118-00-9
EINECS: 285-652-6
UN: 1759
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR