Pentafluorobenzyl bromide ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Purity: 98+%
CAS No.: 1765-40-8
Molecular Formula: C7H2BrF5
Formula Weight: 261.00
Flash Point: 83°(181°F)
Refractive Index: 1.4720
Melting Point: 18-20°
Boiling Point: 174-175°
Beilstein Reference: 647808
UN No.: 3265
MDL No.: MFCD00000299
Vzorec:
(C₆F₅)CH₂Br Bod varu: 174…175 °C Bod tání: 18…20 °C Hustota: 1,844 Bod vzplanutí: 83 °C (181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000299 CAS číslo: 1765-40-8 EINECS: 217-182-4 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...