4-Bromobenzyl chloride ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
4-Bromo-α-chlorotoluene, α-Chloro-4-bromotoluene, 1-Bromo-4-(chloromethyl)benzene, 1-(Chloromethyl)-4-bromobenzene
L11486.06
L11486.14
L11486.06EA
1720
CZK
L11486.06
L11486.14
4-Bromobenzyl chloride ≥97%
4-Bromobenzyl chloride
Purity: 97%
CAS No.: 589-17-3
Molecular Formula: C7H6BrCl
Formula Weight: 205.49
Flash Point:
Vzorec:
C₇H₆BrCl MW: 205,48 g/mol Bod varu: 106…107 °C (10 mmHg) Bod tání: 38…41 °C Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040867 CAS číslo: 589-17-3 EINECS: 209-638-6 UN: 3261 ADR: 8,II Merck Index: 14,1410 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...