Tisk…

α-Bromo-3-chlorotoulene ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: α-Bromo-m-chlorotoulene, 3-Chlorobenzyl bromide, 1-(Bromomethyl)-3-chlorobenzene, alpha-Bromo-3-chlorotoluene

A15844.09 A15844.18
A15844.09EA 1020 CZK
A15844.09 A15844.18
α-Bromo-3-chlorotoulene ≥97%
α-Bromo-3-chlorotoulene

Purity: 97% CAS No.: 766-80-3 Molecular Formula: C7H6BrCl Formula Weight: 205.49 Flash Point: 110°(230°F) Refractive Index: 1.5890 Odour: Pungent Boiling Point: 108-110°/10mm Beilstein Reference: 636504 UN No.: 3265 MDL No.: MFCD00000597

Vzorec: C₇H₆BrCl
MW: 205,48 g/mol
Bod varu: 108…110 °C (10 mmHg)
Bod tání: 17,5 °C
Hustota: 1,567
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000597
CAS číslo: 766-80-3
EINECS: 212-171-0
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR