Tisk…

1-Bromo-4-ethylbenzene ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 4-(Bromophenyl)ethane

A13013.09 A13013.18 A13013.30
A13013.09EA 1530 CZK
A13013.09 A13013.18 A13013.30
1-Bromo-4-ethylbenzene ≥99%
1-Bromo-4-ethylbenzene

Purity: 99% CAS No.: 1585-07-5 Molecular Formula: C8H9Br Formula Weight: 185.06 Flash Point: 63°(145°F) Refractive Index: 1.5440 Boiling Point: 203-204° Beilstein Reference: 1856989 MDL No.: MFCD00000111

Vzorec: C₈H₉Br
Bod varu: 203…204 °C
Hustota: 1,341
Bod vzplanutí: 63 °C (145 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000111
CAS číslo: 1585-07-5
EINECS: 216-439-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR