α,3-Dibromotoluene ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
3-Bromobenzyl bromide, 1-Bromo-3-(bromomethyl)benzene
A15075.09
A15075.18
A15075.30
A15075.09EA
1680
CZK
A15075.09
A15075.18
A15075.30
α,3-Dibromotoluene ≥99%
α,3-Dibromotoluene
Purity: 99%
CAS No.: 823-78-9
Molecular Formula: C7H6Br2
Formula Weight: 249.94
Flash Point:
Vzorec:
C₇H₆Br₂ MW: 249,93 g/mol Bod varu: 128…130 °C (12 mmHg) Bod tání: 40…44 °C Hustota: 1,560 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000176 CAS číslo: 823-78-9 EINECS: 212-519-1 UN: 1759 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...