Tisk…

4-tert-Butyltoluene, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1-(tert-Butyl)-4-methylbenzene

M-Clarity™ quality level = MQ100

B102628-3KG B102628-500G
SIALB102628-3KGEA 12350 CZK
SIALB102628-3KG SIALB102628-500G
4-tert-Butyltoluene, Sigma-Aldrich®
4-tert-Butyltoluene
Vzorec: C₁₁H₁₆
MW: 148,25 g/mol
Bod varu: 192,8 °C (1013 hPa)
Bod tání: –62,5 °C
Hustota: 0,861 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008837
CAS číslo: 98-51-1
EINECS: 202-675-9
UN: 2667
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR