Tisk…

2-Chloro-3-cyano-4-pyridineboronic acid pinacol ester, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Chloro-3-cyanopyridine-4-boronic acid pinacol ester

718920-250MG 718920-1G
SIAL718920-250MGEA 6090 CZK
SIAL718920-250MG SIAL718920-1G
2-Chloro-3-cyano-4-pyridineboronic acid pinacol ester, Sigma-Aldrich®
2-Chloro-3-cyano-4-pyridineboronic acid pinacol ester
Vzorec: C₁₂H₁₄BClN₂O₂
MW: 264,52 g/mol
MDL Number: MFCD09037479
CAS číslo: 878194-94-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR