Tisk…

2-Acetamido-5-pyridineboronic acid pinacol ester, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Acetamidopyridine-5-boronic acid, pinacol ester, N-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)acetamide

M-Clarity™ quality level = MQ100

683892-1G 683892-250MG
SIAL683892-1GEA 6480 CZK
SIAL683892-1G SIAL683892-250MG
2-Acetamido-5-pyridineboronic acid pinacol ester, Sigma-Aldrich®
2-Acetamido-5-pyridineboronic acid pinacol ester
Vzorec: C₁₃H₁₉BN₂O₃
MW: 262,12 g/mol
MDL Number: MFCD08690229
CAS číslo: 904326-87-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR