Tisk…

1-Benzyloxy-3-bromobenzene, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-Benzyloxybromobenzene, 1-(Benzyloxy)-3-bromobenzene

M-Clarity™ quality level = MQ100

668419-10G 668419-50G
SIAL668419-10GEA 2440 CZK
SIAL668419-10G SIAL668419-50G
1-Benzyloxy-3-bromobenzene, Sigma-Aldrich®
1-Benzyloxy-3-bromobenzene
Vzorec: C₁₃H₁₁BrO
MW: 263,13 g/mol
MDL Number: MFCD00155065
CAS číslo: 53087-13-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR