Tisk…

Phenylboronic acid pinacol ester, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, Benzeneboronic acid, pinacol ester, 4,4,5,5-Tetramethyl-2-phenyl-1,3,2-dioxaborolane, 2-Phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

M-Clarity™ quality level = MQ100

647098-10G 647098-50G 647098-5G
SIAL647098-10GEA 6800 CZK
SIAL647098-10G SIAL647098-50G SIAL647098-5G
Phenylboronic acid pinacol ester, Sigma-Aldrich®
Phenylboronic acid pinacol ester
Vzorec: C₁₂H₁₇BO₂
MW: 204,08 g/mol
MDL Number: MFCD00966985
CAS číslo: 24388-23-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR