4-Pyridineboronic acid pinacol ester, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Pyridine-4-boronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
M-Clarity™ quality level = MQ100
578770-1G
578770-5G
SIAL578770-1GEA
6450
CZK
SIAL578770-1G
SIAL578770-5G
4-Pyridineboronic acid pinacol ester, Sigma-Aldrich®
4-Pyridineboronic acid pinacol ester
Vzorec:
C₁₁H₁₆BNO₂ MW: 205,06 g/mol |
MDL Number:
MFCD01319051 CAS číslo: 181219-01-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...