Tisk…

1-Benzyloxy-4-bromobenzene, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Benzyloxybromobenzene, 1-Bromo-4-(phenylmethoxy)benzene

513032-10G 513032-50G
SIAL513032-10GEA 1630 CZK
SIAL513032-10G SIAL513032-50G
1-Benzyloxy-4-bromobenzene, Sigma-Aldrich®
1-Benzyloxy-4-bromobenzene
Vzorec: C₁₃H₁₁BrO
MW: 263,13 g/mol
MDL Number: MFCD00028016
CAS číslo: 6793-92-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR