Tisk…

Methyl-4-pentenoate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyl pent-4-enoate

49661-1G-F 49661-5G-F
SIAL49661-1G-FEA 4070 CZK
SIAL49661-1G-F SIAL49661-5G-F
Methyl-4-pentenoate, Sigma-Aldrich®
Methyl-4-pentenoate
Vzorec: C₆H₁₀O₂
MW: 114,14 g/mol
MDL Number: MFCD03990590
CAS číslo: 818-57-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR