Tisk…

Isobutylboronic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Methylpropylboronic acid, (2-Methylpropyl)boronic acid

M-Clarity™ quality level = MQ200

346225-1G 346225-5G
SIAL346225-1GEA 928 CZK
SIAL346225-1G SIAL346225-5G
Isobutylboronic acid, Sigma-Aldrich®
Isobutylboronic acid
Vzorec: C₄H₁₁BO₂
MDL Number: MFCD00134156
CAS číslo: 84110-40-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR