(5-Fluoro-2-methoxyphenyl)acetic acid
Dodavatel: Apollo Scientific
Synonyms:
2-(5-Fluoro-2-methoxyphenyl)acetic acid
PC7723-1G
PC7723-5G
PC7723-10G
APOSPC7723-1GEA
705
CZK
APOSPC7723-1G
APOSPC7723-5G
APOSPC7723-10G
(5-Fluoro-2-methoxyphenyl)acetic acid
(5-Fluoro-2-methoxyphenyl)acetic acid
Vzorec:
C₉H₉FO₃ MW: 184,17 g/mol |
MDL Number:
MFCD00671771 CAS číslo: 383134-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...