Tisk…

4-Methylphenylboronic acid neopentyl glycol ester ≥99%

Dodavatel: Thermo Fisher Scientific

L19563.03 L19563.06
L19563.03EA 1080 CZK
L19563.03 L19563.06
4-Methylphenylboronic acid neopentyl glycol ester ≥99%
4-Methylphenylboronic acid neopentyl glycol ester

Purity: 99% CAS No.: 380481-66-3 Molecular Formula: C12H17BO2 Formula Weight: 204.08 Melting Point: 93-95° MDL No.: MFCD03788728

Vzorec: C₁₂H₁₇BO₂
Bod tání: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD03788728
CAS číslo: 380481-66-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR