2-Phenylethylboronic acid pinacol ester ≥99%
Dodavatel: Thermo Fisher Scientific
L19530.03
L19530.MD
L19530.03EA
3040
CZK
L19530.03
L19530.MD
2-Phenylethylboronic acid pinacol ester ≥99%
2-Phenylethylboronic acid pinacol ester
Purity: 99%
CAS No.: 165904-22-3
Molecular Formula: C14H21BO2
Formula Weight: 232.13
Refractive Index: 1.4860
Melting Point: 31-33°
Boiling Point: 70-80°
Beilstein Reference: 8258927
MDL No.: MFCD03788721
Vzorec:
C₁₄H₂₁BO₂ Bod varu: 70…80 °C Bod tání: 31…33 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788721 CAS číslo: 165904-22-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...