(2-Chloro-5-(trifluoromethyl)phenyl)acetic acid ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
2-(2-Chloro-5-(trifluoromethyl)phenyl)acetic acid
L19153.03
L19153.06
L19153.03EA
2340
CZK
L19153.03
L19153.06
(2-Chloro-5-(trifluoromethyl)phenyl)acetic acid ≥97%
(2-Chloro-5-(trifluoromethyl)phenyl)acetic acid
Purity: 97%
CAS No.: 22893-39-6
Molecular Formula: C9H6ClF3O2
Formula Weight: 238.59
Melting Point: 112-114°
Beilstein Reference: 2053702
Vzorec:
C₉H₆ClF₃O₂ MW: 238.59 Bod tání: 112…114 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631471 CAS číslo: 22893-39-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...