1-Bromo-2-ethylbenzene ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
B23449.06
B23449.14
B23449.22
B23449.06EA
1170
CZK
B23449.06
B23449.14
B23449.22
1-Bromo-2-ethylbenzene ≥98%
1-Bromo-2-ethylbenzene
Purity: 98%
CAS No.: 1973-22-4
Molecular Formula: C8H9Br
Formula Weight: 185.06
Flash Point: 71°(159°F)
Refractive Index: 1.5485
Melting Point: -68°
Boiling Point: 199-200°
Beilstein Reference: 1929735
MDL No.: MFCD00000077
Vzorec:
C₈H₉Br MW: 185,06 g/mol Bod varu: 199…200 °C Bod tání: –68 °C Hustota: 1,338 Bod vzplanutí: 71 °C (159 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000077 CAS číslo: 1973-22-4 EINECS: 217-823-8 UN: 3082 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...