2-Hydroxy-5-pyridineboronic acid pinacol ester ≥97%
Dodavatel: Thermo Scientific
Synonyms:
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-ol, 6-Hydroxypyridine-3-boronic acid pinacol ester
H51673.MD
H51673.03
H51673.MDEA
824
CZK
H51673.MD
H51673.03
2-Hydroxy-5-pyridineboronic acid pinacol ester ≥97%
2-Hydroxy-5-pyridineboronic acid pinacol ester
Purity: 97%
CAS No.: 1054483-78-1
Molecular Formula: C11H16BNO3
Formula Weight: 221.06
MDL No.: MFCD06795671
Vzorec:
C₁₁H₁₆BNO₃ MW: 221,06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06795671 CAS číslo: 1054483-78-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...